3,5-Dinitrobenzosyre 98+%

Leverandør: Thermo Fisher Scientific
Warning

153191000 153195000 153190010
ACRO153191000EA 485 NOK
ACRO153191000 ACRO153195000 ACRO153190010
3,5-Dinitrobenzosyre 98+%
3,5-Dinitrobenzosyre
Formel: C₇H₄N₂O₆
Smeltepunkt: 205…209 °C
Tetthet: 1,683 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007253
CAS-nummer: 99-34-3
EINECS: 202-751-1
ADR: 4.1,III
Merck Indeks: 13,03304

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 204 to 209 °C
HPLC ≥98.0 %
Titration with NaOH 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR