Giemsa's stain, pure, high purity biological stain

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

153230050 153230250
ACRO153230050EA 465 NOK
ACRO153230050 ACRO153230250
Giemsa's stain, pure, high purity biological stain
Giemsa's stain
Smeltepunkt: 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081642
CAS-nummer: 51811-82-6
EINECS: 257-438-2

Bestill nå

Spesifikasjon Testresultater

Appearance Dark green to black Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤10 % (110°C)
UV Lambda max 520 to 525 nm (in MeOH)
Specific absorbance A (1%/1cm) ≥600 (in MeOH) On dried substance


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR