Quinoline-N-oxide hydrate 98%

Leverandør: Thermo Fisher Scientific

Synonymer: Quinoline 1-oxide hydrate

153430050 153430250
ACRO153430050EA 615 NOK
ACRO153430050 ACRO153430250
Quinoline-N-oxide hydrate 98%
Quinoline-N-oxide hydrate

CAS No.: 1613-37-2

Formel: C₉H₇NO·xH₂O
Smeltepunkt: 48…53 °C
Flash punkt: >112 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149472
CAS-nummer: 198878-42-1

Bestill nå

Spesifikasjon Testresultater

Appearance Ochre to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 48 to 53 °C
Water ≤22.5 % (K.F.)
Titration with HClO4 ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR