2-Chloro-p-toluidine 98%

Leverandør: Thermo Fisher Scientific
Danger

154670250 154670050
ACRO154670250EA 935 NOK
ACRO154670250 ACRO154670050
2-Chloro-p-toluidine 98%
2-Chloro-p-toluidine
Formel: C₇H₈ClN
Kokepunkt: 223…225 °C (760 mmHg)
Smeltepunkt: 7 °C
Tetthet: 1,151 g/cm³
Flash punkt: 99 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007666
CAS-nummer: 615-65-6
EINECS: 210-440-7
FN: 3429
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR