Diethylamine hydrochloride 99%

Leverandør: Thermo Fisher Scientific
Warning

155890010 155891000 155895000 155890051
ACRO155890010EA 1300 NOK
ACRO155890010 ACRO155891000 ACRO155895000 ACRO155890051
Diethylamine hydrochloride 99%
Diethylamine hydrochloride
Formel: C₄H₁₂ClN
Kokepunkt: 320…330 °C (760 mmHg)
Smeltepunkt: 227…230 °C
Tetthet: 1,0475 g/cm³ (21 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012499
CAS-nummer: 660-68-4
EINECS: 211-541-9
Merck Indeks: 13,03138

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 227 to 230 °C
Loss on drying ≤1 %
Titration with HClO4 ≥98.5 %
UV at 420 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR