Glyoxal 40% in aqueous solution

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Ethandial

156220010 156220025 156225000 156220250
ACRO156220010EA 1025 NOK
ACRO156220010 ACRO156220025 ACRO156225000 ACRO156220250
Glyoxal 40% in aqueous solution
Glyoxal
Formel: C₂H₂O₂
MW: 58.04 g/mol
Kokepunkt: 51
Smeltepunkt: –14 °C
Tetthet: 1.265 g/cm³
Flash punkt: >104 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00006957
CAS-nummer: 107-22-2
EINECS: 203-474-9
Merck Indeks: 13,04522

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to yellow Solution (May darken on storage)
Titration excess NaOH 39 to 41 %
Acidity (CH3COOH) ≤0.25 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR