Strychnine 98%

Leverandør: Thermo Fisher Scientific
Danger

158950250 158951000
ACRO158950250EA 1400 NOK
ACRO158950250 ACRO158951000
Strychnine 98%
Strychnine
Formel: C₂₁H₂₂N₂O₂
Smeltepunkt: 284…286 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005941
CAS-nummer: 57-24-9
EINECS: 200-319-7
FN: 1692
ADR: 6.1,I

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
Chloride (Cl) ≤0.01 %
Iron (Fe) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR