Trifenylfosfin sulfid 99+%

Leverandør: Thermo Fisher Scientific
Warning

159030100
ACRO159030100EA 700 NOK
ACRO159030100
Trifenylfosfin sulfid 99+%
Trifenylfosfin sulfid
Formel: C₁₈H₁₅PS
Smeltepunkt: 162…164 °C
Tetthet: 1,191 g/cm³ (25 °C)
MDL nummer: MFCD00004917
CAS-nummer: 3878-45-3
EINECS: 223-407-7
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline solid
Infrared spectrum Conforms
Melting point 162 to 164 °C
GC ≥99.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR