1,3-Dihydroksynaftalen 99+%

Leverandør: Thermo Fisher Scientific
Warning

163340010 163340050 163340250
ACRO163340010EA 945 NOK
ACRO163340010 ACRO163340050 ACRO163340250
1,3-Dihydroksynaftalen 99+%
1,3-Dihydroksynaftalen
Formel: C₁₀H₈O₂
Kokepunkt: 386 °C (1013 hPa)
Smeltepunkt: 123…126 °C
Tetthet: 1,12 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003965
CAS-nummer: 132-86-5
EINECS: 205-079-7
Merck Indeks: 12,06483

Bestill nå

Spesifikasjon Testresultater

Appearance Pink-orange to brown Crystalline powder
Infrared spectrum Conforms
Melting point 123 to 126 °C
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR