Triethoxysilane 95%

Leverandør: Thermo Fisher Scientific
Danger

164810100 164810500 164812500
ACRO164810100EA 860 NOK
ACRO164810100 ACRO164810500 ACRO164812500
Triethoxysilane 95%
Triethoxysilane
Formel: C₆H₁₆O₃Si
Kokepunkt: 134…135 °C (760 mmHg)
Smeltepunkt: –170 °C
Tetthet: 0,89 g/cm³
Flash punkt: 26 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009061
CAS-nummer: 998-30-1
FN: 3489
ADR: 6.1,I

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3770 to 1.3790 (20°C, 589 nm)
GC ≥94 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR