3-Chlorophenylhydrazinium chloride 97%

Leverandør: Thermo Scientific
Warning

Synonymer: 1-(3-Chlorophenyl)hydrazine hydrochloride, 3-Chlorophenylhydrazine hydrochloride

167360250
ACRO167360250EA 1725 NOK
ACRO167360250
3-Chlorophenylhydrazinium chloride 97%
3-Chlorophenylhydrazinium chloride
Formel: C₆H₈Cl₂N₂
MW: 179,05 g/mol
Smeltepunkt: 242 °C (dec.)
MDL nummer: MFCD02656655
CAS-nummer: 2312-23-4
EINECS: 219-005-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to light beige to pink Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR