4-Bromo-m-toluidine 97%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 3-Amino-6-bromotoluene, 4-Bromo-3-methylaniline, 5-Amino-2-bromotoluene

167502500
ACRO167502500EA 3825 NOK
ACRO167502500
4-Bromo-m-toluidine 97%
4-Bromo-m-toluidine
Formel: C₇H₈BrN
MW: 186.05 g/mol
Kokepunkt: 240 °C (760 mmHg)
Smeltepunkt: 79…83 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007828
CAS-nummer: 6933-10-4
EINECS: 230-056-3
FN: 2811
ADR: 6.1 III

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to grey to light brown Powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR