8-Pentadecanone 98%

Leverandør: Thermo Fisher Scientific

169790250
ACRO169790250EA 7975 NOK
ACRO169790250
8-Pentadecanone 98%
8-Pentadecanone
Formel: C₁₅H₃₀O
Kokepunkt: 293 °C (1013 hPa)
Smeltepunkt: 40…44 °C
Tetthet: 0,787 g/cm³ (20 °C)
Flash punkt: 110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009555
CAS-nummer: 818-23-5
EINECS: 212-450-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow Glistening crystals or flakes
Infrared spectrum Conforms
Melting point 40 to 44 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR