Dijodometan 99+% stabilised

Leverandør: Thermo Fisher Scientific
Danger


Stabilisering: stabilised with silver wire

169831000 169835000 169830250 169830025
ACRO169831000EA 1550 NOK
ACRO169831000 ACRO169835000 ACRO169830250 ACRO169830025
Dijodometan 99+% stabilised
Dijodometan
Formel: CH₂I₂
Kokepunkt: 181 °C (760 mmHg)
Smeltepunkt: 6 °C
Tetthet: 3,32 g/cm³
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001079
CAS-nummer: 75-11-6
EINECS: 200-841-5
FN: 2810
ADR: 6.1,III
Merck Indeks: 12,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e37a85a8930c2cbeaf62626d427944a69722cc72&vid=73a1847aa5acf8fa3b0e777fe1bcae9e4f7bde3d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Color) Clear colorless to yellow to red to orange-brown
Appearance (Form) Liquid
Infrared spectrum Conforms
GC ≥99.0 %
Stabilizer Silver


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR