Triethanolamine hydrochloride 99+%

Leverandør: Thermo Fisher Scientific

170051000 170055000 170050025
ACRO170051000EA 640 NOK
ACRO170051000 ACRO170055000 ACRO170050025
Triethanolamine hydrochloride 99+%
Triethanolamine hydrochloride
Formel: C₆H₁₆ClNO₃
Smeltepunkt: 177…179 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012596
CAS-nummer: 637-39-8
EINECS: 211-284-2

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
Loss on drying ≤1.0 % (105°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR