4-Chloroanisole 99%

Leverandør: Thermo Fisher Scientific
Warning

170151000 170150250
ACRO170151000EA 2025 NOK
ACRO170151000 ACRO170150250
4-Chloroanisole 99%
4-Chloroanisole
Formel: C₇H₇ClO
Kokepunkt: 198…202 °C (760 mmHg)
Smeltepunkt: –18 °C
Tetthet: 1,16 g/cm³
Flash punkt: 78 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000616
CAS-nummer: 623-12-1
EINECS: 210-772-2

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation +220° to +235° (20°C, 589 nm) (c=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR