Methoxyacetone 96%

Leverandør: Thermo Fisher Scientific
Warning

175730100 175730500
ACRO175730100EA 1950 NOK
ACRO175730100 ACRO175730500
Methoxyacetone 96%
Methoxyacetone
Formel: C₄H₈O₂
Kokepunkt: 118 °C (1013 hPa)
Smeltepunkt: –66 °C
Tetthet: 0,95 g/cm³
Flash punkt: 25 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008768
CAS-nummer: 5878-19-3
EINECS: 227-549-0
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.4000 (20°C, 589 nm)
GC ≥95 %
Specific optical rotation -63.5° to -67.5° (20°C, 589 nm) (c=1, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR