2'-Fluoroacetophenone 97%

Leverandør: Thermo Fisher Scientific
Warning

179780250
ACRO179780250EA 1800 NOK
ACRO179780250
2'-Fluoroacetophenone 97%
2'-Fluoroacetophenone
Formel: C₈H₇FO
Kokepunkt: 82…83 °C (10 mmHg)
Tetthet: 1,137 g/cm³
Flash punkt: 61 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000320
CAS-nummer: 445-27-2
EINECS: 207-156-0

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5065 to 1.5085 (20°C, 589 nm)
GC ≥96.0 %
Titration with HClO4 99 to 101 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR