2-Methoxyphenethylamine 98%

Leverandør: Thermo Fisher Scientific
Danger

182170250 182171000 182170050
ACRO182170250EA 1600 NOK
ACRO182170250 ACRO182171000 ACRO182170050
2-Methoxyphenethylamine 98%
2-Methoxyphenethylamine
Formel: C₉H₁₃NO
Kokepunkt: 236…237 °C
Tetthet: 1,03 g/cm³
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008186
CAS-nummer: 2045-79-6
EINECS: 218-066-6
FN: 2735
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5400 to 1.5420 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR