α,α,α-Trifluoro-p-toluic acid 98%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 4-(Trifluoromethyl)benzoic acid

188460010 188460050
ACRO188460010EA 340 NOK
ACRO188460010 ACRO188460050
α,α,α-Trifluoro-p-toluic acid 98%
α,α,α-Trifluoro-p-toluic acid
Formel: C₈H₅F₃O₂
MW: 190.12 g/mol
Smeltepunkt: 219…222 °C
MDL nummer: MFCD00002562
CAS-nummer: 455-24-3
EINECS: 207-242-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to light grey Crystalline powder
Infrared spectrum Conforms
Melting point 219 to 222 °C
GC ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR