Erythrosin B disodium salt, pure, high purity biological stain

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: C.I. 45430

189460250
ACRO189460250EA 795 NOK
ACRO189460250
Erythrosin B disodium salt, pure, high purity biological stain
Erythrosin B disodium salt
Formel: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00144257
CAS-nummer: 16423-68-0
EINECS: 240-474-8

Bestill nå

Spesifikasjon Testresultater

Appearance Red to bordeaux Powder
Infrared spectrum Conforms
Dye content ≥87 %
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Lead (Pb) ≤10 ppm
Mercury (Hg) ≤1 ppm
Insoluble matter ≤0.2 % (in water)
Subsidiary dyes ≤4.0 %
Ether extractable matter ≤0.2 %
Fluorescein ≤20 ppm
Inorganic iodide ≤0.1 % (as Sodium iodide)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR