Resazurin sodium salt, pure, high purity biological stain

Leverandør: Thermo Fisher Scientific
Warning

189900010 189900050 189900250
ACRO189900010EA 570 NOK
ACRO189900010 ACRO189900050 ACRO189900250
Resazurin sodium salt, pure, high purity biological stain
Resazurin sodium salt
Formel: C₁₂H₆NNaO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005036
CAS-nummer: 62758-13-8
EINECS: 263-718-5

Bestill nå

Spesifikasjon Testresultater

Appearance Dark green to black Powder
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥75 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR