Carmine alum lake of carminic acid, pure, high purity stain_biological

Leverandør: Thermo Fisher Scientific

Synonymer: CI 75470

190200050 190200250 190201000
ACRO190200050EA 1050 NOK
ACRO190200050 ACRO190200250 ACRO190201000
Carmine alum lake of carminic acid, pure, high purity stain_biological
Carmine
Formel: C₂₂H₂₀O₁₃
MW: 492.389 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167028
CAS-nummer: 1390-65-4
EINECS: 215-724-4

Bestill nå

Spesifikasjon Testresultater

Appearance Dark red to purple Powder
Infrared spectrum Conforms
UV spectrum Complies
Assay ≥42.0 % (Carminic acid)
Loss on drying ≤20 %
Ash ≤12 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR