2-Bromo-6-methoxynaphthalene 98%

Leverandør: Thermo Fisher Scientific
Warning

190560050 190560250
ACRO190560050EA 495 NOK
ACRO190560050 ACRO190560250
2-Bromo-6-methoxynaphthalene 98%
2-Bromo-6-methoxynaphthalene
Formel: C₁₁H₉BrO
Smeltepunkt: 105…111 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004062
CAS-nummer: 5111-65-9
EINECS: 225-837-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 105 to 111 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR