Ethyl cellulose, 10 cps 48% (ethoxyl content) 10 cps

Leverandør: Thermo Fisher Scientific

190962500 190965000
ACRO190962500EA 1525 NOK
ACRO190962500 ACRO190965000
Ethyl cellulose, 10 cps 48% (ethoxyl content) 10 cps
Ethyl cellulose

Appearance: White to light yellow Crystalline powder or granules

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00131037
CAS-nummer: 9004-57-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Powder or granules
Infrared spectrum Conforms
Assay 47.5 to 49.5 % (Ethoxyl content)
Viscosity 8 to 12 cP (5% in 80/20 toluene/ethanol, 25°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR