Ethyl cellulose, 22 cps 48% (ethoxyl content)

Leverandør: Thermo Fisher Scientific

190972500 190975000 190970025
ACRO190972500EA 1425 NOK
ACRO190972500 ACRO190975000 ACRO190970025
Ethyl cellulose, 22 cps 48% (ethoxyl content)
Ethyl cellulose

CAS No.: 9004-57-3

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00131037
CAS-nummer: 9004-57-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow Crystalline powder or granules
Infrared spectrum Conforms
Assay 48 to 49.5 % (Ethoxyl)
Viscosity 18 to 24 cP (5% soln. in 80/20 toluene/ethanol, 25°C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR