Cesium acetate 99%, ren

Leverandør: Thermo Fisher Scientific
Warning

191040250 191041000
ACRO191040250EA 1025 NOK
ACRO191040250 ACRO191041000
Cesium acetate 99%, ren
Cesium acetate
Formel: H₃CCOOCs
Smeltepunkt: 194 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013056
CAS-nummer: 3396-11-0
EINECS: 222-248-0

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder and/or chunks
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR