Antimony(III) fluoride 98%

Leverandør: Thermo Fisher Scientific
Danger

191241000 191245000
ACRO191241000EA 760 NOK
ACRO191241000 ACRO191245000
Antimony(III) fluoride 98%
Antimony(III) fluoride
Formel: F₃Sb
Kokepunkt: 376 °C (1013 hPa)
Smeltepunkt: 292 °C
Tetthet: 4,38 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011218
CAS-nummer: 7783-56-4
EINECS: 232-009-2
FN: 2923
ADR: 8,II
Merck Indeks: 13,00715

Bestill nå

Spesifikasjon Testresultater

Appearance White to grey Crystalline powder
Titration Iodimetric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR