Rhodamine 6G, pure

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

191410500
ACRO191410500EA 865 NOK
ACRO191410500
Rhodamine 6G, pure
Rhodamine 6G
Formel: C₂₈H₃₁ClN₂O₃
Kokepunkt: ∼200 °C (1013 hPa)
MDL nummer: MFCD00012665
CAS-nummer: 989-38-8
EINECS: 213-584-9
FN: 2811

Bestill nå

Spesifikasjon Testresultater

Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR