Magnesiumnitrat heksahydrat 99.999% (trace metals basis)

Leverandør: Thermo Fisher Scientific

193420100 193420500
ACRO193420100EA 670 NOK
ACRO193420100 ACRO193420500
Magnesiumnitrat heksahydrat 99.999% (trace metals basis)
Magnesiumnitrat heksahydrat
Formel: Mg(NO₃)₂·6H₂O
Kokepunkt: 330 °C (760 mmHg)
Smeltepunkt: 89 °C
Tetthet: 1,63 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149779
CAS-nummer: 13446-18-9
ADR: 5.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder or crystals and/or chunks
Titration Complexometric 9.1 to 9.7 % (Mg)
Total trace metal impurities ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR