Palladium (II) oksid 99.995% (trace metals basis)

Leverandør: Thermo Fisher Scientific

193680010 193680050
ACRO193680010EA 4550 NOK
ACRO193680010 ACRO193680050
Palladium (II) oksid 99.995% (trace metals basis)
Palladium (II) oksid
Formel: OPd
Smeltepunkt: 870 °C (dec.)
Tetthet: 8,73 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011172
CAS-nummer: 1314-08-5

Bestill nå

Spesifikasjon Testresultater

Appearance Grey to black Powder or granules and/or chunks
Assay 85.2 to 88.7 % (Pd)
Total trace metal impurities ≤50 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR