Natriumsulfat, vannfri 99+% for HPLC

Leverandør: Thermo Fisher Scientific

194100500 194102500
ACRO194100500EA 825 NOK
ACRO194100500 ACRO194102500
Natriumsulfat, vannfri 99+% for HPLC
Sodium sulphate
Formel: Na₂SO₄
Kokepunkt: >1700 °C (760 mmHg)
Smeltepunkt: 884 °C
Tetthet: 2,7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003504
CAS-nummer: 7757-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Water ≤0.1 %
UV (10 % in water)
Acidity/alkalinity ≤0.0002 meq/g
Titration with HClO4 ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR