Hafnium 1.000 mg/l Hf in 5% hydrofluoric acid standard til AAS
Leverandør: Thermo Fisher Scientific
196011000
ACRO196011000EA 720 NOK
ACRO196011000
Hafnium 1.000 mg/l Hf in 5% hydrofluoric acid standard til AAS
Standarder Kationstandarder Hafnium Standards