Kalsiumhydrid ∼ 93%, grains 0-20 mm, extra pure (up to 15% powder)

Leverandør: Thermo Fisher Scientific
Danger

196791000
ACRO196791000EA 1625 NOK
ACRO196791000
Kalsiumhydrid ∼ 93%, grains 0-20 mm, extra pure (up to 15% powder)
Kalsiumhydrid
Formel: H₂Ca
Smeltepunkt: 675 °C
Tetthet: 1,9 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010897
CAS-nummer: 7789-78-8
EINECS: 232-189-2
FN: 1404
ADR: 4.3,I
Merck Indeks: 13,01674

Bestill nå

Spesifikasjon Testresultater

Appearance Grey Granules
Assay ≥92 % (Total Ca)
Particle size 1 to 20 mm
Aluminium (Al) ≤100 ppm
Hydrogen (H) 980 ml/g CaH2
Iron (Fe) ≤100 ppm
Magnesium (Mg) ≤8000 ppm
Nitrogen (N) ≤2000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR