kobolt (II) acetat tetrahydrat 98-102% ACS

Leverandør: Thermo Fisher Scientific
Danger

197101000
ACRO197101000EA 1225 NOK
ACRO197101000
kobolt (II) acetat tetrahydrat 98-102% ACS
kobolt (II) acetat tetrahydrat
Formel: (H₃CCOO)₂Co·4H₂O
Smeltepunkt: 140 °C
Tetthet: 1,7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149569
CAS-nummer: 6147-53-1
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance Pink to red Crystalline powder or granules
Titration Complexometric 98.0 to 102.0 %
Chloride (Cl) ≤0.002 %
Nitrate (NO3) ≤0.01 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.002 %
Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.001 %
Magnesium (Mg) ≤0.005 %
Nickel (Ni) ≤0.1 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.05 %
Zinc (Zn) ≤0.01 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR