Molekylsil 3A (0,3 nm, 3 Å) 4 --> 8 mesh

Leverandør: Thermo Fisher Scientific

197240050 197245000
ACRO197240050EA 8775 NOK
ACRO197240050 ACRO197245000
Molekylsil 3A (0,3 nm, 3 Å) 4 --> 8 mesh
Molekylsil 3A (0,3 nm, 3 Å)
Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL nummer: MFCD00131613
CAS-nummer: 308080-99-1

Bestill nå

Spesifikasjon Testresultater

Appearance Beige to tan Beads
Particle size 4 to 8 mesh
Bulk density ≥600 g/l
Residual water ≤1.5 %
Water adsorption ≥20 % (60% rH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR