Ruthenium(IV) oxide hydrate (> 54% Ru)

Leverandør: Thermo Fisher Scientific
Warning

197430010 197430050 197430250
ACRO197430010EA 1025 NOK
ACRO197430010 ACRO197430050 ACRO197430250
Ruthenium(IV) oxide hydrate (> 54% Ru)
Ruthenium(IV) oxide hydrate
Formel: O₂Ru·H₂O
Tetthet: 6,97 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149846
CAS-nummer: 32740-79-7
EINECS: 234-840-6

Bestill nå

Spesifikasjon Testresultater

Appearance Black Powder
Assay ≥54 % (Ru)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR