Kobber(II)sulfat pentahydrat 98%, extra pure

Leverandør: Thermo Fisher Scientific
Danger

197730010 197730050 197730250
ACRO197730010EA 720 NOK
ACRO197730010 ACRO197730050 ACRO197730250
Kobber(II)sulfat pentahydrat 98%, extra pure
Kobber(II)sulfat pentahydrat
Formel: CuSO₄·5H₂O
Smeltepunkt: 110 °C (dec.)
Tetthet: 2,284 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149681
CAS-nummer: 7758-99-8
EINECS: 231-847-6
FN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Indeks: 13,02682

Bestill nå

Spesifikasjon Testresultater

Appearance Blue Crystalline powder or crystals
Titration Iodometric ≥97.5 %
Arsenic (As) ≤10 ppm
Iron (Fe) ≤200 ppm
Lead (Pb) ≤125 ppm
Nickel (Ni) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR