4-Hexylresorcinol 98%

Leverandør: Thermo Fisher Scientific
Warning

197920250 197921000
ACRO197920250EA 790 NOK
ACRO197920250 ACRO197921000
4-Hexylresorcinol 98%
4-Hexylresorcinol
Formel: C₁₂H₁₈O₂
Kokepunkt: 333 °C (1013 hPa)
Smeltepunkt: 65…70 °C
Flash punkt: ≥ 100 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002284
CAS-nummer: 136-77-6
EINECS: 205-257-4
Merck Indeks: 12,04750

Bestill nå

Spesifikasjon Testresultater

Appearance White to light pink to light orange Powder and/or chunks
Infrared spectrum Conforms
Melting point 63 to 68 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR