Natriumtetrahydroborat 99%, pulver

Leverandør: Thermo Fisher Scientific
Danger

200051000 200050250 200055000
ACRO200051000EA 1950 NOK
ACRO200051000 ACRO200050250 ACRO200055000
Natriumtetrahydroborat 99%, pulver
Sodium borohydride
Formel: NaBH₄
Smeltepunkt: 360 °C (dec.)
Tetthet: 1,07 g/cm³ (20 °C)
Flash punkt: 69 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003518
CAS-nummer: 16940-66-2
EINECS: 241-004-4
FN: 1426
ADR: 4.3,I
Merck Indeks: 13,08664

Bestill nå

Spesifikasjon Testresultater

Appearance White Powder
Titration Iodometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR