Iminodiacetic acid 98+%

Leverandør: Thermo Fisher Scientific
Warning

204970250 204975000 204970025
ACRO204970250EA 380 NOK
ACRO204970250 ACRO204975000 ACRO204970025
Iminodiacetic acid 98+%
Iminodiacetic acid
Formel: C₄H₇NO₄
Smeltepunkt: 243 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004280
CAS-nummer: 142-73-4
EINECS: 205-555-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR