Formamid 99.5% for analysis

Leverandør: Thermo Fisher Scientific
Danger

205820010 205820025 205821000 205820100
ACRO205820010EA 1075 NOK
ACRO205820010 ACRO205820025 ACRO205821000 ACRO205820100
Formamid 99.5% for analysis
Formamid
Formel: HCONH₂
Kokepunkt: 210 °C (760 mmHg)
Smeltepunkt: 2…3 °C
Tetthet: 1,133 g/cm³
Flash punkt: 175 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007941
CAS-nummer: 75-12-7
EINECS: 200-842-0
Merck Indeks: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.05 % (Coulometric)
Sulfated ash ≤0.005 %
GC ≥99.5 %
Color scale ≤10 APHA
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Zinc (Zn) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR