1-Heptansulfonsyre natriumsalt 98%

Leverandør: Thermo Fisher Scientific
Warning

206130050 206130250 206131000 206135000
ACRO206130050EA 755 NOK
ACRO206130050 ACRO206130250 ACRO206131000 ACRO206135000
1-Heptansulfonsyre natriumsalt 98%
1-Heptansulfonsyre natriumsalt
Formel: H₃C(CH₂)₅CH₂SO₃Na
Smeltepunkt: >300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007543
CAS-nummer: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Powder or flakes
Infrared spectrum Conforms
Loss on drying ≤9 % (100°C, 3 h) (vacuum)
Solubility (10 % in water) Clear colorless
Titration after Ion exchange ≥97.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR