di-Litiumtetraborat 98+% for analysis

Leverandør: Thermo Fisher Scientific
Danger

206940010 206942500
ACRO206940010EA 5750 NOK
ACRO206940010 ACRO206942500
di-Litiumtetraborat 98+% for analysis
di-Litiumtetraborat
Formel: Li₂B₄O₇
Smeltepunkt: 930 °C
Tetthet: 1,4 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011083
CAS-nummer: 12007-60-2
EINECS: 234-514-3
Merck Indeks: 14,05524

Bestill nå

Spesifikasjon Testresultater

Appearance White Powder
Loss on drying ≤0.5 % (1 g, 200°C)
Titration with HCl ≥98.0 %
Chloride (Cl) ≤50 ppm
Phosphate (PO4) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR