CO₂ absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Ascarite(II)®

Leverandør: Thermo Fisher Scientific
208085000 208080025 208081000
ACRO208085000EA 5700 NOK
ACRO208085000 ACRO208080025 ACRO208081000
CO₂ absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Ascarite(II)®
Eksikatorer Tørkemidler
CAS No.: 81133-20-2

MDL No.: MFCD00132264
UN: 1823
Hazard Class: 8
Packing group: II
GHS Symbols: GHS05
Appearance: Brown Granules
Storage Temperature: Ambient
Signal word: Danger
Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR