Platinium(IV) oxide hydrate Pt 79-84%

Leverandør: Thermo Fisher Scientific
Danger

211920010 211920050 211920250
ACRO211920010EA 2575 NOK
ACRO211920010 ACRO211920050 ACRO211920250
Platinium(IV) oxide hydrate Pt 79-84%
Platinium(IV) oxide hydrate
Formel: O₂Pt·H₂O
Smeltepunkt: 450 °C
Tetthet: 10,2 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066964
CAS-nummer: 52785-06-5
EINECS: 215-223-0
FN: 1479
ADR: 5.1,I
Merck Indeks: 12,07686

Bestill nå

Spesifikasjon Testresultater

Appearance Brown to black Powder
Assay 79 to 84 % (Pt)
Surface area ≥90 m²/g (BET)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR