Spermidine trihydrochloride 99+%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 4-Azoniaoctamethylenediammonium trichloride, 4-Azaoctamethylenediamine trihydrochloride, N1-(3-Aminopropyl)butane-1,4-diamine trihydrochloride, N-(3-Aminopropyl)-1,4-diaminobutane trihydrochloride

215100050 215100010 215100250
ACRO215100050EA 1775 NOK
ACRO215100050 ACRO215100010 ACRO215100250
Spermidine trihydrochloride 99+%
Spermidine trihydrochloride
Formel: C₇H₂₂Cl₃N₃
MW: 254.63 g/mol
Smeltepunkt: 248…252 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00012918
CAS-nummer: 334-50-9
EINECS: 206-379-0
Merck Indeks: 12,08893

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR