Ammoniumacetat 98+% for HPLC

Leverandør: Thermo Fisher Scientific

219990500 219992500
ACRO219990500EA 1300 NOK
ACRO219990500 ACRO219992500
Ammoniumacetat 98+% for HPLC
Ammoniumacetat
Formel: H₃CCOONH₄
Smeltepunkt: 114 °C
Tetthet: 1,17 g/cm³ (20 °C)
Flash punkt: 136 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013066
CAS-nummer: 631-61-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Powder or crystals
UV at 280 nm A: ≤0.02
pH 6.5 to 7.5 (5% soln.in water)
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR