3-Iodoanisole 97%

Leverandør: Thermo Fisher Scientific
Warning

222020050 222020250 222021000
ACRO222020050EA 455 NOK
ACRO222020050 ACRO222020250 ACRO222021000
3-Iodoanisole 97%
3-Iodoanisole
Formel: C₇H₇IO
Kokepunkt: 244…245 °C (760 mmHg)
Tetthet: 1,96 g/cm³
Flash punkt: >110 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001048
CAS-nummer: 766-85-8
EINECS: 212-173-1

Bestill nå

Spesifikasjon Testresultater

Appearance Clear yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6130 to 1.6150 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR