Magnesiumoksid 97+% for analysis

Leverandør: Thermo Fisher Scientific

222532500 222530010
ACRO222532500EA 1125 NOK
ACRO222532500 ACRO222530010
Magnesiumoksid 97+% for analysis
Magnesiumoksid
Formel: MgO
Kokepunkt: 3600 °C (760 mmHg)
Smeltepunkt: 2800 °C
Tetthet: 3,58 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011109
CAS-nummer: 1309-48-4
EINECS: 215-171-9
Merck Indeks: 13,05700

Bestill nå

Spesifikasjon Testresultater

Appearance White Powder
Titration Complexometric ≥97.0 % (on dry substance)
Chloride (Cl) ≤0.1 %
Sulfate (SO4) ≤1.0 %
Calcium (Ca) ≤1.1 %
Iron (Fe) ≤0.05 %
Heavy metals (as Pb) ≤0.003 %
Soluble matter ≤2 % (in water)
Insoluble matter ≤0.1 % (in HCl)
Loss on ignition ≤5.0 % (900°C)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR